N,N'-Bis(3-aminopropyl)ethylenediamine ≥95%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
1,5,8,12-Tetraazadodecane
07-1961-100G
07-1961-500G
07-1961-25G
100208-274EA
63.69
USD
100208-274
100201-352
100201-350
N,N'-Bis(3-aminopropyl)ethylenediamine ≥95%
N,N'-Bis(3-aminopropyl)ethylenediamine
Formula:
C₈H₂₂N₄ MW: 174.29 g/mol Boiling Pt: 118 °C (0.2 mmHg) Density: 0,952 Flash Pt: >230 °F |
MDL Number:
MFCD00008210 CAS Number: 10563-26-5 UN: 2927 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...