1,4,8,11-Tetraazacyclotetradecane ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
07-1959-1G
07-1959-250MG
07-1959-5G
100206-242EA
104.74
USD
100206-242
100198-504
100209-548
1,4,8,11-Tetraazacyclotetradecane ≥98%
1,4,8,11-Tetraazacyclotetradecane
Formula:
C₁₀H₂₄N₄ MW: 200.33 g/mol Melting Pt: 185…186 °C |
MDL Number:
MFCD00005105 CAS Number: 295-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...