1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
DOTA, 1,4,7,10-Tetraazacyclododecane-N,N',N'',N'''-tetraacetic acid, 2,2',2'',2'''-(1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetrayl)tetraacetic acid
07-1942-1G
07-1942-250MG
07-1942-5G
100208-764EA
447.26
USD
100208-764
100208-762
100208-766
1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid ≥98%
1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid
MOF
Formula:
C₁₆H₂₈N₄O₈ MW: 404.42 g/mol |
MDL Number:
MFCD00068657 CAS Number: 60239-18-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...