1-Benzyl-1,4,7,10-tetraazacyclododecane ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: N-Benzyl-1,4,7,10-tetraazacyclododecane

07-1939-250MG 07-1939-1G
101442-398EA 302.89 USD
101442-398 101442-400
1-Benzyl-1,4,7,10-tetraazacyclododecane ≥98%
1-Benzyl-1,4,7,10-tetraazacyclododecane
Formula: C₁₅H₂₆N₄
MW: 262.401 g/mol
MDL Number: MFCD09263310
CAS Number: 112193-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR