Pyridinium trifluoromethanesulfonate ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Pyridinium Triflate
07-1885-5G
07-1885-25G
100209-546EA
60.86
USD
100209-546
100209-544
Pyridinium trifluoromethanesulfonate ≥97%
Pyridinium trifluoromethanesulfonate
Formula:
C₆H₆F₃NO₃S MW: 229.17 g/mol Melting Pt: 221…223 °C |
MDL Number:
MFCD00274656 CAS Number: 52193-54-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...