Protoporphyrin IX, dimethyl ester

Supplier: STREM CHEMICALS INC MS

Synonyms: Dimethyl 3,3'-(3,8,13,17-tetramethyl-7,12-divinylporphyrin-2,18-diyl)dipropionate

07-1850-100MG 07-1850-500MG
100209-542EA 185.42 USD
100209-542 100210-780
Protoporphyrin IX, dimethyl ester
Protoporphyrin IX, dimethyl ester
Formula: C₃₆H₃₈N₄O₄
MW: 590.7113 g/mol
MDL Number: MFCD00064531
CAS Number: 5522-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR