Poly(4-vinylpyridine) (cross-linked), anhydrous, beads, (toluene conditioned)
Supplier: STREM CHEMICALS INC MS
07-1765-25G
07-1765-500G
07-1765-100G
100199-324EA
392.06
USD
100199-324
100207-332
100202-736
Poly(4-vinylpyridine) (cross-linked), anhydrous, beads, (toluene conditioned)
Poly(4-vinylpyridine) (cross-linked)
Formula:
[-CH₂CH(C₅H₄N)CH₂-]n Boiling Pt: 225 °C/ 260 °C (dec. air/ dec. N₂) Density: 1.15 |
MDL Number:
MFCD00190588 CAS Number: 9017-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...