2,3,7,8,12,13,17,18-Octaethylporphyrin 97

Supplier: STREM CHEMICALS INC MS

Synonyms: OEP, Octaethylporphine, 2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine

07-1550-1G 07-1550-250MG
100209-294EA 1002.09 USD
100209-294 100199-724
2,3,7,8,12,13,17,18-Octaethylporphyrin 97
2,3,7,8,12,13,17,18-Octaethylporphyrin

Porphine

Formula: C₃₆H₄₆N₄
MW: 534.79 g/mol
MDL Number: MFCD00009627
CAS Number: 2683-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR