Mesoporphyrin IX dihydrochloride

Supplier: STREM CHEMICALS INC MS

07-1100-10MG 07-1100-50MG
100195-982EA 69.35 USD
100195-982 100202-868
Mesoporphyrin IX dihydrochloride
Mesoporphyrin IX dihydrochloride
Formula: C₂₄H₃₈N₄O₄.2HCl
MW: 639.61 g/mol
MDL Number: MFCD00136049
CAS Number: 68938-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR