1,3-Dimesitylimidazol-2-ylidene ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: 1,3-Bis(2,4,6-trimethylphenyl)imidazol-2-ylidene, IMes, 1,3-Bis(2,4,6-trimethylphenyl)-1,3-dihydro-2H-imidazol-2-ylidene

07-0600-2G 07-0600-500MG
101418-422EA 441.6 USD
101418-422 101418-420
1,3-Dimesitylimidazol-2-ylidene ≥98%
1,3-Dimesitylimidazol-2-ylidene

NHC, Organocatalyst

Formula: C₂₁H₂₄N₂
MW: 304.44 g/mol
MDL Number: MFCD09265338
CAS Number: 141556-42-5
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR