1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride ≥97%

Supplier: STREM CHEMICALS INC MS

07-0590-500MG 07-0590-2G
100208-754EA 123.14 USD
100208-754 100208-756
1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride ≥97%
1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride

NHC

Formula: C₂₇H₃₇ClN₂
MDL Number: MFCD02684545
CAS Number: 250285-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR