meso-1,2-Bis(naphthyl)ethylenediamine ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

07-0492-500MG 07-0492-100MG
101442-384EA 324.12 USD
101442-384 101442-382
meso-1,2-Bis(naphthyl)ethylenediamine ≥98%
meso-1,2-Bis(naphthyl)ethylenediamine
MDL Number: MFCD05150370
CAS Number: 117106-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR