1,3-Diisopropylimidazolium chloride ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 1,3-Di-i-propylimidazolium chloride, 1,3-Diisopropyl-1H-imidazol-3-ium chloride

07-0485-1G 07-0485-5G
100206-238EA 100.49 USD
100206-238 100206-240
1,3-Diisopropylimidazolium chloride ≥97%
1,3-Diisopropylimidazolium chloride

NHC

Formula: C₉H₁₇ClN₂
MW: 188.70 g/mol
MDL Number: MFCD03840577
CAS Number: 139143-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR