(11bS)-4,4-Dibutyl-2,6-bis(3,4,5-trifluorophenyl)-4,5-dihydro-3H-dinaphtho[2,1-c:1',2'-e]azepinium bromide

Supplier: STREM CHEMICALS INC MS
Warning

07-0381-50MG 07-0381-250MG
101418-396EA 356.68 USD
101418-396 101418-398
(11bS)-4,4-Dibutyl-2,6-bis(3,4,5-trifluorophenyl)-4,5-dihydro-3H-dinaphtho[2,1-c:1',2'-e]azepinium bromide
(11bS)-4,4-Dibutyl-2,6-bis(3,4,5-trifluorophenyl)-4,5-dihydro-3H-dinaphtho[2,1-c:1',2'-e]azepinium bromide

Organocatalyst

MDL Number: MFCD09264271
CAS Number: 851942-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR