(S)-(-)-2,2'-Diamino-1,1'-binaphthalene ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S)-1,1'-Binaphthyl-2,2'-diamine, (S)-(-)-BINAM, (S)-(-)-1,1'-Binaphthyl-2,2'-diamine, (S)-(-)-1,1'-Bi(2-naphthylamine), (S)-(-)-2,2'-Diamino-1,1'-binaphthyl

07-0328-1G 07-0328-250MG
100203-898EA 312.8 USD
100203-898 100206-228
(S)-(-)-2,2'-Diamino-1,1'-binaphthalene ≥99%
(S)-(-)-2,2'-Diamino-1,1'-binaphthalene
Formula: C₂₀H₁₆N₂
MW: 284.362 g/mol
Melting Pt: 242…244 °C
MDL Number: MFCD00145204
CAS Number: 18531-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR