1,1'-Binaphthyl-2,2'-diamine ≥96%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: BINAM, [1,1'-Binaphthalene]-2,2'-diamine, racemic-2,2'-Diamino-1,1'-binaphthyl, 2,2'-Diamino-1,1'-binaphthyl, (±)-1,1'-Bi(2-naphthylamine)

07-0326-5G 07-0326-1G
100208-262EA 1406.89 USD
100208-262 100208-260
1,1'-Binaphthyl-2,2'-diamine ≥96%
1,1'-Binaphthyl-2,2'-diamine
Formula: C₂₀H₁₆N₂
MW: 284.36 g/mol
MDL Number: MFCD00145204
CAS Number: 4488-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR