Deuteroporphyrin IX, dimethyl ester ≥97%

Supplier: STREM CHEMICALS INC MS

07-0325-10MG 07-0325-50MG
100201-344EA 67.94 USD
100201-344 100195-968
Deuteroporphyrin IX, dimethyl ester ≥97%
Deuteroporphyrin IX, dimethyl ester
Melting Pt: 227 °C
MDL Number: MFCD00010033
CAS Number: 10589-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR