Coproporphyrin I tetramethyl ester synthetic 98

Supplier: STREM CHEMICALS INC MS

07-0310-10MG 07-0310-50MG
100199-720EA 133.05 USD
100199-720 100210-604
Coproporphyrin I tetramethyl ester synthetic 98
Coproporphyrin I tetramethyl ester
Melting Pt: 251…252 °C
MDL Number: MFCD00079038
CAS Number: 25767-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR