(S,S)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (-)-2,6-Bis[(4S)-4-(i-propyl)-2-oxazolin-2-yl]pyridine, (S,S)-2,2'-(2,6-Pyridinediyl)bis(4-isopropyl-2-oxazoline), 2,6-Bis((S)-4-isopropyl-4,5-dihydrooxazol-2-yl)pyridine, (S)-(i-Pr)-pybox

07-0307-250MG 07-0307-1G
101418-386EA 217.97 USD
101418-386 101418-388
(S,S)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine ≥98%
(S,S)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine

Pincer Ligands and Complexes, Pybox

Formula: C₁₇H₂₃N₃O₂
Melting Pt: 152…153 °C
MDL Number: MFCD00191815
CAS Number: 118949-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR