(R,R)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
2,6-Bis((R)-4-isopropyl-4,5-dihydrooxazol-2-yl)pyridine, (+)-2,6-Bis[(4R)-4-(i-propyl)-2-oxazolin-2-yl]pyridine, (R)-(i-Pr)-pybox, (R,R)-2,2'-(2,6-Pyridinediyl)bis(4-isopropyl-2-oxazoline)
07-0306-250MG
07-0306-1G
101418-382EA
208.06
USD
101418-382
101418-384
(R,R)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine ≥98%
(R,R)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine
Pincer Ligands and Complexes, Pybox
Formula:
C₁₇H₂₃N₃O₂ MW: 301.39 g/mol Melting Pt: 152…153 °C |
MDL Number:
MFCD01321334 CAS Number: 131864-67-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...