2,6-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: (S,S)-Ph-pybox, (-)-2,6-Bis[(4S)-4-phenyl-2-oxazolin-2-yl]pyridine

07-0304-2G 07-0304-500MG
100203-704EA 750.15 USD
100203-704 100199-718
2,6-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine ≥98%
2,6-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine

Pincer Ligands and Complexes, Pybox

Formula: C₂₃H₁₉N₃O₂
Melting Pt: 171…175 °C
MDL Number: MFCD01863586
CAS Number: 174500-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR