2,6-Bis((R)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: (R,R)-Ph-pybox, (+)-2,6-Bis[(4R)-4-phenyl-2-oxazolin-2-yl]pyridine

07-0303-500MG 07-0303-2G
100206-220EA 242.03 USD
100206-220 100202-852
2,6-Bis((R)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine ≥98%
2,6-Bis((R)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine

Pincer Ligands and Complexes, Pybox

Formula: C₂₃H₁₉N₃O₂
MW: 369.42 g/mol
Melting Pt: 171…175 °C
MDL Number: MFCD01863585
CAS Number: 128249-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR