1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride ≥97%
Supplier: STREM CHEMICALS INC MS
Synonyms:
1,3-Dimesityl-1H-imidazol-3-ium chloride, 1,3-Dimesitylimidazolium chloride
07-0299-1G
07-0299-5G
100209-520EA
97.66
USD
100209-520
100209-522
1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride ≥97%
1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride
NHC
Formula:
C₂₁H₂₅ClN₂ |
MDL Number:
MFCD02684541 CAS Number: 141556-45-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...