N,N'-Bis(2-hydroxybenzyl)ethylenediamine ≥98%

Supplier: STREM CHEMICALS INC MS

07-0295-25G 07-0295-5G
100205-914EA 716.18 USD
100205-914 100202-850
N,N'-Bis(2-hydroxybenzyl)ethylenediamine ≥98%
N,N'-Bis(2-hydroxybenzyl)ethylenediamine
Formula: C₁₆H₂₀N₂O₂
Melting Pt: 118…120 °C
MDL Number: MFCD00058922
CAS Number: 18653-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR