(1R,2R)-(+)-1,2-Bis(4-nitrophenyl)ethylenediamine dihydrochloride ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

07-0243-500MG 07-0243-100MG
101418-378EA 677.97 USD
101418-378 101418-376
(1R,2R)-(+)-1,2-Bis(4-nitrophenyl)ethylenediamine dihydrochloride ≥98%
(1R,2R)-(+)-1,2-Bis(4-nitrophenyl)ethylenediamine dihydrochloride
MDL Number: MFCD09834838
CAS Number: 117903-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR