(1R,2R)-(-)-1,2-Bis(4-hydroxyphenyl)ethylenediamine dihydrochloride ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

07-0232-100MG 07-0232-500MG
101418-372EA 172.68 USD
101418-372 101418-374
(1R,2R)-(-)-1,2-Bis(4-hydroxyphenyl)ethylenediamine dihydrochloride ≥98%
(1R,2R)-(-)-1,2-Bis(4-hydroxyphenyl)ethylenediamine dihydrochloride
MDL Number: MFCD09834837
CAS Number: 1055301-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR