(1R,2R)-(+)-1,2-Bis(4-dimethylaminophenyl)ethylenediamine tetrahydrochloride ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

07-0226-100MG 07-0226-500MG
101418-368EA 165.6 USD
101418-368 101418-370
(1R,2R)-(+)-1,2-Bis(4-dimethylaminophenyl)ethylenediamine tetrahydrochloride ≥98%
(1R,2R)-(+)-1,2-Bis(4-dimethylaminophenyl)ethylenediamine tetrahydrochloride
MDL Number: MFCD09839134
CAS Number: 866267-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR