(1R,2R)-(+)-1,2-Bis(4-cyanophenyl)ethylenediamine dihydrochloride ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

07-0220-100MG 07-0220-500MG
101418-364EA 167.02 USD
101418-364 101418-366
(1R,2R)-(+)-1,2-Bis(4-cyanophenyl)ethylenediamine dihydrochloride ≥98%
(1R,2R)-(+)-1,2-Bis(4-cyanophenyl)ethylenediamine dihydrochloride
MDL Number: MFCD09834836
CAS Number: 117903-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR