(4S,5R)-(-)-4-Benzyl-5-(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)-2-oxazolidinone ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

07-0095-500MG 07-0095-100MG
101433-046EA 587.38 USD
101433-046 101433-044
(4S,5R)-(-)-4-Benzyl-5-(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)-2-oxazolidinone ≥99%
(4S,5R)-(-)-4-Benzyl-5-(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)-2-oxazolidinone
Melting Pt: 90…95 °C
MDL Number: MFCD09834832
CAS Number: 857637-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR