neo-Decanoic acid, Prime grade
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Neodecanoic acid
06-5000-100G
06-5000-500G
100201-336EA
58.03
USD
100201-336
100204-476
neo-Decanoic acid, Prime grade
neo-Decanoic acid
Formula:
C₁₀H₂₀O₂ MW: 172.2700 g/mol Boiling Pt: 243…253 °C Density: 0.92 |
MDL Number:
MFCD00068291 CAS Number: 26896-20-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...