Potassium tricyanomethanide ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
06-1700-1G
06-1700-5G
100207-522EA
167.02
USD
100207-522
100209-290
Potassium tricyanomethanide ≥98%
Potassium tricyanomethanide
Formula:
C₄KN₃ |
MDL Number:
MFCD00058850 CAS Number: 34171-69-2 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...