(+)-1,4-Di-O-tosyl-2,3-O-isopropylidene-D-threitol

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (4R,5R)-(+)-4,5-Bis(tosyloxymethyl)-2,2-dimethyl-1,3-dioxolane, ((4R,5R)-2,2-Dimethyl-1,3-dioxolane-4,5-diyl)bis(methylene) bis(4-methylbenzenesulphonate), (4R,5R)-(+)-2,3-O-Isopropylidene-1,4-di-O-tosyl-D-threitol, (4R,5R)-(-)-O-Isopropylidene-2,3-dihydroxy-1,4-bis(p-tosyl)butane, (4R,5R)-(+)-2,2-Dimethyl-4,5-bis(tosyloxymethyl)-1,3-dioxide

06-1050-1G 06-1050-5G
100204-786EA 141.54 USD
100204-786 100201-332
(+)-1,4-Di-O-tosyl-2,3-O-isopropylidene-D-threitol
(+)-1,4-Di-O-tosyl-2,3-O-isopropylidene-D-threitol
Formula: C₂₁H₂₆O₈S₂
MW: 470.55 g/mol
Melting Pt: 89…91 °C
MDL Number: MFCD00063235
CAS Number: 51064-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR