(-)-1,4-Di-O-tosyl-2,3-O-isopropylidene-L-threitol
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
((4S,5S)-2,2-Dimethyl-1,3-dioxolane-4,5-diyl)bis(methylene) bis(4-methylbenzenesulphonate), (4S,5S)-(-)-2,3-O-Isopropylidene-1,4-di-O-tosyl-L-threitol, (4S,5S)-(+)-O-Isopropylidene-2,3-dihydroxy-1,4-bis(p-tosyl)butane
06-1040-5G
06-1040-1G
100202-732EA
553.42
USD
100202-732
100204-784
(-)-1,4-Di-O-tosyl-2,3-O-isopropylidene-L-threitol
(-)-1,4-Di-O-tosyl-2,3-O-isopropylidene-L-threitol
Formula:
C₂₁H₂₆O₈S₂ MW: 470.55 g/mol Melting Pt: 89…91 °C |
MDL Number:
MFCD00003212 CAS Number: 37002-45-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...