Fullerenes - C60/C70 mixture (contains ~20% C70 and ~1% higher fullerenes)

Supplier: STREM CHEMICALS INC MS
Warning

06-0500-1G 06-0500-250MG 06-0500-50MG
100206-210EA 464.25 USD
100206-210 100206-208 100204-778
Fullerenes - C60/C70 mixture (contains ~20% C70 and ~1% higher fullerenes)
Fullerenes - C60/C70 mixture

Fullerene

Formula: C60, C70
Density: 1.6
MDL Number: MFCD00282904
CAS Number: 131159-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR