Carbon nanotube multi-walled ≥85% (nanotubes), Length ~1-5 µm, Diameter ~20-25 nm

Supplier: STREM CHEMICALS INC MS
Warning

06-0475-1G 06-0475-250MG
100202-480EA 656.74 USD
100202-480 100208-256
Carbon nanotube multi-walled ≥85% (nanotubes), Length ~1-5 µm, Diameter ~20-25 nm
Carbon nanotube

Carbon Nanotubes, Nanomaterials

Formula: C
MDL Number: MFCD00133992
CAS Number: 308068-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR