racemic-4-Bromo[2.2]paracyclophane ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 4-Bromo[2.2]paracyclophane

06-0104-2G 06-0104-500MG
101418-354EA 247.69 USD
101418-354 101418-352
racemic-4-Bromo[2.2]paracyclophane ≥95%
racemic-4-Bromo[2.2]paracyclophane

Paracyclophane

Formula: C₁₆H₁₅Br
MW: 287.1943 g/mol
MDL Number: MFCD00187358
CAS Number: 1908-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR