Triphenylmethylium tetrakis(pentafluorophenyl)borate ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Trityltetra(pentafluorophenyl)borate, Triphenylmethylium tetrakis(perfluorophenyl)borate, Trityl tetrakis(pentafluorophenyl)borate, Triphenylcarbenium Tetrakis(pentafluorophenyl)borate, Tritylium Tetrakis(pentafluorophenyl)borate
05-5000-5G
05-5000-1G
100198-496EA
791.2
USD
100198-496
100198-494
Triphenylmethylium tetrakis(pentafluorophenyl)borate ≥97%
Triphenylmethylium tetrakis(pentafluorophenyl)borate
Formula:
C₄₃H₁₅BF₂₀ MW: 922.37 g/mol |
MDL Number:
MFCD03426981 CAS Number: 136040-19-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...