Tris(pentafluorophenyl)borane ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Tris(perfluorophenyl)borane
05-1500-25G
05-1500-1G
05-1500-5G
100198-488EA
1932
USD
100198-488
100198-486
100198-490
Tris(pentafluorophenyl)borane ≥97%
Tris(pentafluorophenyl)borane
Metallocenes, Derivatives & Cp Precursors
Formula:
C₁₈BF₁₅ MW: 511.98 g/mol |
MDL Number:
MFCD00269813 CAS Number: 1109-15-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...