Triphenylborane, ammonia complex ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

05-1295-5G 05-1295-1G
100206-200EA 785.54 USD
100206-200 100206-198
Triphenylborane, ammonia complex ≥98%
Triphenylborane, ammonia complex
MDL Number: MFCD03788909
CAS Number: 13276-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR