1H-Pyrazole-4-boronic acid pinacol ester ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

05-1048-1G 05-1048-5G
100210-940EA 142.95 USD
100210-940 100210-942
1H-Pyrazole-4-boronic acid pinacol ester ≥97%
1H-Pyrazole-4-boronic acid pinacol ester

Boronic Acid and Ester

Formula: C₉H₁₅BN₂O₂
MDL Number: MFCD03453063
CAS Number: 269410-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR