4-Hydroxyphenylboronic acid pinacol ester ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 4-Hydroxybenzeneboronic acid pinacol ester

05-1044-5G 05-1044-1G
100202-600EA 529.35 USD
100202-600 100202-598
4-Hydroxyphenylboronic acid pinacol ester ≥97%
4-Hydroxyphenylboronic acid pinacol ester

Boronic Acid and Ester

Formula: C₁₂H₁₇BO₃
Melting Pt: 114…116 °C
MDL Number: MFCD02093756
CAS Number: 269409-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR