Furan-3-boronic acid pinacol ester ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
2-(3-Furanyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)furan
05-1039-1G
05-1039-5G
100204-470EA
110.4
USD
100204-470
100204-472
Furan-3-boronic acid pinacol ester ≥97%
Furan-3-boronic acid pinacol ester
Boronic Acid and Ester
Formula:
C₁₀H₁₅BO₃ MW: 194.035 g/mol Melting Pt: 63…64 °C |
MDL Number:
MFCD03453669 CAS Number: 248924-59-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...