(4-Acetamidophenyl)boronic acid pinacol ester ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)acetanilide, 4-Acetylaminophenylboronic acid pinacol ester, N-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetamide
05-1032-1G
05-1032-5G
100195-944EA
130.22
USD
100195-944
100195-946
(4-Acetamidophenyl)boronic acid pinacol ester ≥97%
(4-Acetamidophenyl)boronic acid pinacol ester
Boronic Acid and Ester
Formula:
C₁₄H₂₀BNO₃ MW: 261.125 g/mol Melting Pt: 165…167 °C |
MDL Number:
MFCD02093722 CAS Number: 214360-60-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...