4-Hydroxy-3-methoxyphenylboronic acid pinacol ester ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 2-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

05-0950-5G 05-0950-1G
100210-934EA 829.42 USD
100210-934 100210-932
4-Hydroxy-3-methoxyphenylboronic acid pinacol ester ≥97%
4-Hydroxy-3-methoxyphenylboronic acid pinacol ester

Boronic Acid and Ester

Formula: C₁₃H₁₉BO₄
MW: 250.099 g/mol
Melting Pt: 104…107 °C
MDL Number: MFCD02093723
CAS Number: 269410-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR