3-(Ethoxycarbonyl)phenylboronic acid pinacol ester ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Ethyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 3-Ethoxycarbonylphenylboronic acid, pinacol ester, Ethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

05-0775-5G 05-0775-1G
100203-692EA 533.6 USD
100203-692 100203-690
3-(Ethoxycarbonyl)phenylboronic acid pinacol ester ≥97%
3-(Ethoxycarbonyl)phenylboronic acid pinacol ester

Boronic Acid and Ester

Formula: C₁₅H₂₁BO₄
MW: 276.136 g/mol
Melting Pt: 41…42 °C
MDL Number: MFCD03093896
CAS Number: 269410-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR