4-(Ethoxycarbonyl)phenylboronic acid pinacol ester ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 4-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, Ethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

05-0765-5G 05-0765-1G
100201-310EA 560.49 USD
100201-310 100201-308
4-(Ethoxycarbonyl)phenylboronic acid pinacol ester ≥97%
4-(Ethoxycarbonyl)phenylboronic acid pinacol ester

Boronic Acid and Ester

Formula: C₁₅H₂₁BO₄
Boiling Pt: 105…115 °C (0.01 mmHg)
Density: 1,052
MDL Number: MFCD03453659
CAS Number: 195062-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR