2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 3,5-Dimethyl-4-hydroxyphenylboronic acid pinacol ester, 3,5-Dimethy-4-hydroxyphenylboronic acid pinacol ester

05-0760-1G 05-0760-5G
100202-594EA 111.82 USD
100202-594 100202-596
2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥97%
3,5-Dimethyl-4-hydroxyphenylboronic acid pinacol ester

Boronic Acid and Ester

Formula: C₁₄H₂₁BO₃
MW: 248.13 g/mol
Melting Pt: 105…109 °C
MDL Number: MFCD02093724
CAS Number: 269410-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR