N,N-Dimethylanilinium tetrakis(pentafluorophenyl)borate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Dimethylanilinium tetrakis (pentafluorophenyl)borate, N,N-Dimethylanilinium tetra(pentafluorophenyl)borate, N,N-Dimethylbenzenaminium tetrakis(perfluorophenyl)borate

05-0750-5G 05-0750-1G
100208-614EA 946.89 USD
100208-614 100208-612
N,N-Dimethylanilinium tetrakis(pentafluorophenyl)borate ≥98%
N,N-Dimethylanilinium tetrakis(pentafluorophenyl)borate

Metallocenes, Derivatives & Cp Precursors

Formula: C₃₂H₁₂BF₂₀N
MW: 801.23 g/mol
MDL Number: MFCD12913898
CAS Number: 118612-00-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR