3-Carboxyphenylboronic acid ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 3-Boronobenzoic acid, 3-Carboxybenzeneboronic acid

05-0399-1G 05-0399-5G
100197-038EA 94.83 USD
100197-038 100197-040
3-Carboxyphenylboronic acid ≥97%
3-Carboxyphenylboronic acid

Boronic Acid and Ester

Formula: C₇H₇BO₄
MDL Number: MFCD00036833
CAS Number: 25487-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR