t-Butyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl carbonate ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

05-0380-1G 05-0380-5G
100198-062EA 94.83 USD
100198-062 100198-064
t-Butyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl carbonate ≥97%
t-Butyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl carbonate

Boronic Acid and Ester

Melting Pt: 86…88 °C
MDL Number: MFCD03427273
CAS Number: 480438-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR